Merck
Investigation of fusion gene expression in HCT116 cells.
Oncology letters (2017-11-29)
Yanmei Zhang, Juan Ren, Mengdie Fang, Xiaoju Wang
ABSTRACT
Colon cancer is the most common type of gastrointestinal cancer. A number of specific and sensitive biomarkers facilitate the diagnosis and monitoring of patients with colon cancer. Fusion genes are typically identified in cancer and a majority of the newly identified fusion genes are oncogenic in nature. Therefore, fusion genes are potential biomarkers and/or therapy targets in cancer. In the present study, the regulation of specific candidate fusion genes were investigated using Brother of the Regulator of Imprinted Sites (BORIS) in the HCT116 colon cancer cell line, which is a paralog of the fusion gene regulator CCCTC-binding factor (CTCF). The copy number of BORIS increased correspondingly to the progression of colorectal carcinoma from the M0 to the M1a stage. It was identified that
Sigma-Aldrich
MISSION® esiRNA, targeting human CTCFL
Pricing and availability is not currently available.