Merck
  • Purification process for MUFA- and PUFA-based monoacylglycerols from edible oils.
Purification process for MUFA- and PUFA-based monoacylglycerols from edible oils.
Biochimie (2017-06-10)
M J González-Fernández, R P Ramos-Bueno, I Rodríguez-García, J L Guil-Guerrero
ABSTRACT
Important health benefits have been attributed to monoacylglycerols (MAGs) due to their various physiological functions, owing to which they become candidates for use as functional foods in order to prevent the onset of certain diseases such as colon cancer. In this work, six edible oils, namely: olive, linseed, sunflower, evening primrose, DHASCO
Product Description
Supelco
PUFA No.1, Marine source, analytical standard