Merck
  • Unusual reactivity of dithiol protected clusters in comparison to monothiol protected clusters: studies using Ag
Unusual reactivity of dithiol protected clusters in comparison to monothiol protected clusters: studies using Ag
Nanoscale (2016-12-03)
Atanu Ghosh, Debasmita Ghosh, Esma Khatun, Papri Chakraborty, Thalappil Pradeep
ABSTRACT
We report the synthesis and unique reactivity of a new green dithiol protected cluster (DTPC), Ag
Sigma-Aldrich
2,4-Dimethylbenzenethiol, 95%
Sigma-Aldrich
1,3-Benzenedithiol, 99%
Sigma-Aldrich
1-Octanethiol, ≥98.5%