Merck
  • Inhibition of Protein Tyrosine Phosphatase 1B Improves IGF-I Receptor Signaling and Protects Against Inflammation-Induced Gliosis in the Retina.
Inhibition of Protein Tyrosine Phosphatase 1B Improves IGF-I Receptor Signaling and Protects Against Inflammation-Induced Gliosis in the Retina.
Investigative ophthalmology & visual science (2016-01-01)
Ana I Arroba, Ángela M Valverde
ABSTRACT
Insulin-like growth factor-I receptor (IGF-IR) signaling mediates retinal growth and survival and its failure may contribute to aggravate diabetic retinopathy (DR). Protein tyrosine phosphatase 1B (PTP1B) negatively modulates IGF-IR signaling, but its involvement in inflammation during DR remains unknown. We investigated whether PTP1B participates in the cross-talk between proinflammatory signaling pathways and IGF-IR-mediated signaling in the retina. 661W photoreceptors or mouse retinal explants were treated with TNFα, IL6, and IL1β. Insulin-like growth factor-I receptor signaling cascade was evaluated in the absence or presence of PTP1B. db/db mice were used to test a PTP1B inhibitor in retinal gliosis. 661W retinal cells and retinal explants responded to IGF-I by inducing IGF-IR tyrosine (13-fold) and Akt phosphorylations (7- and 3-fold for serine 473 and threonine 308, respectively). Cytokines triggered early activation of stress kinases (c-jun [NH2] terminal kinase [JNK] and p38 MAPK), resulting in insulin receptor substrate 1 (IRS1) serine 307 phosphorylation that precedes its degradation. Pretreatment of 661W cells or retinal explants with cytokines upregulated PTP1B protein levels (1.45- and 4.5-fold, respectively), induced IRS1 degradation and decreased IGF-I-mediated IGF-IR/Akt phosphorylation. Silencing or deficiency in PTP1B ameliorated the negative effects of cytokines on IGF-IR signaling. Cytokines increased glial fibrillary acidic protein (GFAP) expression in retinal explants by 4.5-fold, this response being reduced by 2-fold with a PTP1B inhibitor. Protein tyrosine phosphatase 1B protein levels increased by 3-fold in retinas from db/db mice and its inhibition reduced gliosis. Targeting PTP1B might be useful for modulating IGF-I effects in retinal cells during DR.
Sigma-Aldrich
Anti-phospho-IRS1 (Ser307 mouse/ Ser312 human) Antibody, clone 24.6.2, clone 24.6.2, from mouse
Sigma-Aldrich
PTP Assay Kit 1, PTP Assay Kit 1 can be used to detect PTP-1B activity by dephosphorylation of the phosphopeptide (RRLIEDAEpYAARG) using malachite green detection, or by hydrolysis of pNPP.
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
PTP1B Inhibitor, PTP1B Inhibitor, CAS 765317-72-4, is a cell-permeable, selective, reversible and non-competitive allosteric inhibitor of PTP1B (IC₅₀ = 4 µM and 8 µM for PTP1B403 and PTP1B298, respectively).