Merck
An intronic microRNA links Rb/E2F and EGFR signaling.
PLoS genetics (2014-07-25)
Mary Truscott, Abul B M M K Islam, James Lightfoot, Núria López-Bigas, Maxim V Frolov
ABSTRACT
The importance of microRNAs in the regulation of various aspects of biology and disease is well recognized. However, what remains largely unappreciated is that a significant number of miRNAs are embedded within and are often co-expressed with protein-coding host genes. Such a configuration raises the possibility of a functional interaction between a miRNA and the gene it resides in. This is exemplified by the Drosophila melanogaster dE2f1 gene that harbors two miRNAs, mir-11 and mir-998, within its last intron. miR-11 was demonstrated to limit the proapoptotic function of dE2F1 by repressing cell death genes that are directly regulated by dE2F1, however the biological role of miR-998 was unknown. Here we show that one of the functions of miR-998 is to suppress dE2F1-dependent cell death specifically in rbf mutants by elevating EGFR signaling. Mechanistically, miR-998 operates by repressing dCbl, a negative regulator of EGFR signaling. Significantly, dCbl is a critical target of miR-998 since dCbl phenocopies the effects of miR-998 on dE2f1-dependent apoptosis in rbf mutants. Importantly, this regulation is conserved, as the miR-998 seed family member miR-29 repressed c-Cbl, and enhanced MAPK activity and wound healing in mammalian cells. Therefore, the two intronic miRNAs embedded in the dE2f1 gene limit the apoptotic function of dE2f1, but operate in different contexts and act through distinct mechanisms. These results also illustrate that examining an intronic miRNA in the context of its host's function can be valuable in elucidating the biological function of the miRNA, and provide new information about the regulation of the host gene itself.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid