Merck
  • Pharmacokinetic/pharmacodynamic evaluation of the efficacy of flumequine in treating colibacillosis in turkeys.
Pharmacokinetic/pharmacodynamic evaluation of the efficacy of flumequine in treating colibacillosis in turkeys.
Poultry science (2013-11-16)
C Ferraresi, L Lucatello, V Meucci, L Intorre, G Grilli, A Piccirillo, E Russo, R Villa, C Montesissa, P Cagnardi
ABSTRACT
Flumequine (FLU) is used in the treatment of systemic bacterial infections in poultry, including colibacillosis, which is a common disease in turkeys. The pharmacokinetic (PK) behavior of FLU administered to 32 healthy turkeys as an oral bolus via gavage or as 10-h pulsed administration in drinking water were compared, using the authorized dose of 15 mg/kg and the double dose of 30 mg/kg. The minimum inhibitory concentrations (MIC) of 235 Escherichia coli field strains isolated from poultry were determined for pharmacodynamics (PD) to develop a PK/PD model. Blood samples were collected at established times over 24 h, and the obtained plasma was analyzed using a liquid chromatography tandem mass spectrometry method that was validated in-house. A monocompartmental model and a noncompartmental model were applied to the data to obtain the PK results. For both types of administration and both dosages, the ratios of the maximum concentration (Cmax)/MIC50 and the area under the plasma concentration-time curve (AUC)/MIC50 achieved were considerably lower than the fluoroquinolone breakpoints usually adopted for efficacy. The Cmax/MIC50 and AUC0-24/MIC50 ratios were, respectively, 0.67 ± 0.09 and 4.76 ± 0.48 and 1.18 ± 0.35 and 7.05 ± 2.40 for the 15 and 30 mg/kg bolus doses, respectively. After 10-h pulsed administration of 15 mg/kg, values of Cmax/MIC50, 0.19 ± 0.02 on d 1 and 0.30 ± 0.08 on d 5 of therapy were obtained, the AUC/MIC50 ratios were 2.09 ± 0.29 and 3.22 ± 0.93 on d 1 and 5, respectively. Higher values were obtained with the doubled dose of 30 mg/kg: the Cmax/MIC50 ratios were 0.49 ± 0.11 on d 1 and 0.69 ± 0.18 on d 5; the AUC/MIC50 ratios were 5.15 ± 1.15 and 6.57 ± 1.92 on d 1 and 5, respectively. Based on these results, FLU administration should be adopted when specific diagnostic findings indicate its efficacy, and revising the dosage scheme to comply with the prudent and responsible use of antimicrobials in veterinary medicine is advisable.
Product Description
Supelco
Flumequine, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Flumequine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.