Merck
  • Recognition of (2S)-aminomalonyl-acyl carrier protein (ACP) and (2R)-hydroxymalonyl-ACP by acyltransferases in zwittermicin A biosynthesis.
Recognition of (2S)-aminomalonyl-acyl carrier protein (ACP) and (2R)-hydroxymalonyl-ACP by acyltransferases in zwittermicin A biosynthesis.
Biochemistry (2010-04-01)
Yolande A Chan, Michael G Thomas
ABSTRACT
Polyketide synthases elongate a polyketide backbone by condensing carboxylic acid precursors that are thioesterified to either coenzyme A or an acyl carrier protein (ACP). Two of the three known ACP-linked extender units, (2S)-aminomalonyl-ACP and (2R)-hydroxymalonyl-ACP, are found in the biosynthesis of the agriculturally important antibiotic zwittermicin A. We previously reconstituted the formation of (2S)-aminomalonyl-ACP and (2R)-hydroxymalonyl-ACP from the primary metabolites l-serine and 1,3-bisphospho-d-glycerate. In this report, we characterize the two acyltransferases involved in the specific transfer of the (2S)-aminomalonyl and (2R)-hydroxymalonyl moieties from the ACPs associated with extender unit formation to the ACPs integrated into the polyketide synthase. This work establishes which acyltransferase recognizes each extender unit and also provides insight into the substrate selectivity of these enzymes. These are important step toward harnessing these rare polyketide synthase extender units for combinatorial biosynthesis.
Sigma-Aldrich
Tartronic acid, ≥97.0%