Merck
[Chemical constituents from the rhizoma of Arundina graminifolia].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-02-22)
Mei-feng Liu, Yun Han, Dong-ming Xing, Wei Wang, Li-zhen Xu, Li-jun Du, Yi Ding
ABSTRACT
To isolate and elucidate the chemical constituents from the tuber of Arundina graminifolia. The compounds were extracted by 95% alcohol and isolated by column chromatography on silica gel, SephedaxLH-20 and ODS. The structures were determined by UV, IR, NMR and MS spectral analysis. Five compounds were isolated, and their structures were identified as (2E)-, 2-propenoic acid, 3-(4-hydroxy-3-methoxyphenyl)-decosyl ester (I), p-hydroxybenzyl alcohol (II), triacontanol (III) and p-hydroxybenzylethyl ether (IV), 3-hydroxy-5-methoxybibenzyl (V), respectively. All compounds were isolated from the genus of Arundina for the first time.
Sigma-Aldrich
1-Triacontanol, ≥98% (capillary GC)