Merck
Degradation of 4-chlorophenylacetic acid by a Pseudomonas species.
Journal of bacteriology (1981-04-01)
U Klages, A Markus, F Lingens
ABSTRACT
Pseudomonas sp. strain CBS3 was able to utilize 4-chlorophenylacetic acid as the sole source of carbon and energy. When this strain was grown with 4-chlorophenylacetic acid, homoprotocatechuic acid was found to be an intermediate which was further metabolized by the meta-cleavage pathway. Furthermore, three isomers of chlorohydroxyphenylacetic acid, two of them identified as 3-chloro-4-hydroxyphenylacetic acid and 4-chloro-3-hydroxyphenylacetic acid, were isolated from the culture medium. 4-Hydroxyphenylacetic acid was catabolized in a different manner by the glutathione-dependent homogentisate pathway. Degradation enzymes of both of these pathways were inducible.
Sigma-Aldrich
4-Chlorophenylacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
3-Chloro-4-hydroxyphenylacetic acid, 99%