Merck
  • High-performance nanopapers based on benzenesulfonic functionalized graphenes.
High-performance nanopapers based on benzenesulfonic functionalized graphenes.
ACS nano (2012-10-27)
Wenyi Huang, Xilian Ouyang, L James Lee
ABSTRACT
High-performance graphene nanopapers are prepared from an aqueous solution of functional graphenes with benzenesulfonic acid groups via covalent bonds. The formed hydrophobic graphene nanopapers showed the highest tensile strength of 360 MPa and Young's modulus of 102 GPa for samples with 13.7 wt % functional group and annealed at 150 °C. These samples showed a high electrical conductivity of 4.45 × 10(4) S/m after being annealed at 250 °C. The aforementioned properties of graphene nanopapers are much higher than any previously reported data. The properties of nanopapers depend on the degree of functionality on graphenes and the annealing temperatures, which are further evidenced by X-ray photoelectron spectroscopy, FTIR, and X-ray diffraction patterns. Such unique nanopapers can be easily bounded and sandwiched onto any solid surface to give rise to great potentials in many applications such as gas diffusion barriers, EMI shielding, thermal management, and anticorrosion.
Sigma-Aldrich
Sodium benzenesulfonate, 97%
Sigma-Aldrich
Benzenesulfonic acid, technical grade, 90%
Sigma-Aldrich
Benzenesulfonic acid, 98.0% (T)