Merck
  • Etanercept downregulates the Th17 pathway and decreases the IL-17+/IL-10+ cell ratio in patients with psoriasis vulgaris.
Etanercept downregulates the Th17 pathway and decreases the IL-17+/IL-10+ cell ratio in patients with psoriasis vulgaris.
Journal of clinical immunology (2012-06-16)
Emiliano Antiga, Walter Volpi, Elisa Cardilicchia, Laura Maggi, Lucia Filì, Cinzia Manuelli, Paola Parronchi, Paolo Fabbri, Marzia Caproni
ABSTRACT
To evaluate circulating and lesional CD4(+) and CD8(+) cells belonging to Th1, Th2, and Th17 patterns as well as IL-10(+) cells before and after a 12-week lasting course with etanercept or acitretin in patients with psoriasis. 15 patients were given etanercept 50 mg twice weekly and 15 patients acitretin 0,4 mg/kg/day, both for 12 weeks. At the baseline and at the end of the treatment, blood and skin samples were taken to investigate IL-4, IL-8, IL-10, IL-17, and IFN-γ-producing CD4(+) and CD8(+) cells. As controls, 10 healthy controls (HC) and 6 atopic dermatitis (AD) patients were included into the study. Psoriasis patients showed augmented IL-17- and IL-8-producing CD4(+) cells in the blood than HC and AD patients. In the skin lesions, IL-17(+) cells were more represented in psoriasis than in AD, while the number of IL-4-producing cells was reduced in psoriasis patients than in AD ones. Etanercept was able to significantly reduce the number of IL-17- and IL-8-producing CD4(+) and CD8(+) cells both in skin and blood, as well as to augment the proportion of IL-10-producing CD4(+) cells in the skin of psoriatic patients, while acitretin was not. Our results confirmed the role of Th17 cells in the pathogenesis of psoriasis. Etanercept, but not acitretin, was able to downregulate the Th17 pathway and to increase the percentages of IL-10-producing cells in the skin.
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Acitretin, European Pharmacopoeia (EP) Reference Standard