Merck
Immunoelectrophoresis: a method with many faces.
Methods in molecular biology (Clifton, N.J.) (2012-05-16)
Gyorgy Csako
ABSTRACT
Immunoelectrophoresis (IEP) was the first practical method that combined electrophoresis and -immunoprecipitation for identifying and characterizing proteins within complex mixtures. Over the years, IEP has been extended to include a variety of techniques and, as a general name, has been applied to virtually any technique that involves electrophoresis and antigen-antibody precipitin reaction for proteins. Because of the diversity in technical details of different IEP versions, the method described here deals only with classic IEP. Although it requires some manual expertise, IEP is versatile, relatively easy to customize, and economical with no need for expensive instrumentation. Further, it can discern identity, partial identity, and nonidentity of the proteins. Any low-viscosity body fluid specimen or, possibly, culture fluid and tissue extract could be tested with IEP if proper antibodies are available. With these attributes, classic IEP remains a valuable tool for clinical diagnostic testing, purity checking of biochemical and pharmaceutical products, and research.
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis