Merck
  • Azure B, a metabolite of methylene blue, is a high-potency, reversible inhibitor of monoamine oxidase.
Azure B, a metabolite of methylene blue, is a high-potency, reversible inhibitor of monoamine oxidase.
Toxicology and applied pharmacology (2011-12-27)
Anél Petzer, Brian H Harvey, Gregers Wegener, Jacobus P Petzer
ABSTRACT
Methylene blue (MB) has been shown to act at multiple cellular and molecular targets and as a result possesses diverse medical applications. Among these is a high potency reversible inhibition of monoamine oxidase A (MAO-A) that may, at least in part, underlie its adverse effects but also its psycho- and neuromodulatory actions. MB is metabolized to yield N-demethylated products of which azure B, the monodemethyl species, is the major metabolite. Similar to MB, azure B also displays a variety of biological activities and may therefore contribute to the pharmacological profile of MB. Based on these observations, the present study examines the interactions of azure B with recombinant human MAO-A and -B. The results show that azure B is a potent MAO-A inhibitor (IC₅₀=11 nM), approximately 6-fold more potent than is MB (IC₅₀=70 nM) under identical conditions. Measurements of the time-dependency of inhibition suggest that the interaction of azure B with MAO-A is reversible. Azure B also reversibly inhibits the MAO-B isozyme with an IC₅₀ value of 968 nM. These results suggest that azure B may be a hitherto under recognized contributor to the pharmacology and toxicology of MB by blocking central and peripheral MAO-A activity and as such needs to be considered during its use in humans and animals.
Sigma-Aldrich
Azure B, prepared by direct synthesis
Sigma-Aldrich
Azure B, certified by the Biological Stain Commission, certified by the BSC