Merck
  • Chemical markers in Origanum vulgare L. from Kumaon Himalayas: a chemosystematic study.
Chemical markers in Origanum vulgare L. from Kumaon Himalayas: a chemosystematic study.
Natural product research (2011-08-06)
C Pande, G Tewari, S Singh, C Singh
ABSTRACT
The essential oils of four wild growing Origanum vulgare L. (family Lamiaceae) collected from different locations in Kumaon region (Uttarakhand, India) were analysed by capillary GC and GC/MS. The comparative results of O. vulgare L. collected from four different regions showed differences in the chemical constituents of the essential oils. The oil of O. vulgare L. collected from Dhoulchina and Champawat (chemotype I) shows p-cymene (6.7-9.8%), γ-terpinene (12.4-14.0%), thymol (29.7-35.1%) and carvacrol (12.4-20.9%) as major constituents while the oil from Kilbury and Rushi village (chemotype II) shows linalool (6.7-9.7%), bornyl acetate (12.6-16.8%), β-caryophyllene (10.5-13.8%) and germacrene D (6.3-11.3%) as the major constituents. These features highlight the chemosystematics of this genus.
Sigma-Aldrich
γ-Terpinene, ≥95%, FG
Sigma-Aldrich
α-Terpinene, 85%
Sigma-Aldrich
γ-Terpinene, 97%
Sigma-Aldrich
α-Terpinene, ≥95.0% (GC)
Supelco
γ-Terpinene, analytical standard
Sigma-Aldrich
γ-Terpinene, ≥95.0% (GC)
Sigma-Aldrich
α-Terpinene, ≥89%, FCC, FG
Sigma-Aldrich
γ-Terpinene, natural, 95%, FG