Merck
  • All-plasmonic modulation via stimulated emission of copropagating surface plasmon polaritons on a substrate with gain.
All-plasmonic modulation via stimulated emission of copropagating surface plasmon polaritons on a substrate with gain.
Nano letters (2011-05-06)
Alexey V Krasavin, Thanh Phong Vo, Wayne Dickson, Pádraig M Bolger, Anatoly V Zayats
ABSTRACT
We experimentally demonstrate suppressed absorption and stimulated emission of surface plasmon polaritons (SPP) leading to all-plasmonic modulation of an SPP signal propagating at the interface between a metal and a gain medium; these observations are supported by the developed theory. The use of copropagating signal and control waves can provide more than 10 times more efficient SPP stimulated emission compared to out-of-plane pumping and opens up the possibility to realize integratable plasmonic components for active nanophotonic circuitry.
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O