Merck
  • Directed evolution and characterization of a novel D-pantonohydrolase from Fusarium moniliforme.
Directed evolution and characterization of a novel D-pantonohydrolase from Fusarium moniliforme.
Journal of agricultural and food chemistry (2006-08-03)
Zhiqiang Liu, Zhihao Sun, Yong Leng
ABSTRACT
D-Pantonohydrolase has attracted increasing attention as a biocatalyst for stereospecific production of D-pantoic acid. The Fusarium moniliforme D-pantonohydrolase was selected for directed evolution through error-prone Polymerase Chain Reaction (PCR) combined with DNA shuffling for improved activity and pH stability using a convenient two-step high-throughput screening method based on the product formation and pH indicator. After three sequential error-prone PCRs and two rounds of DNA shuffling followed by screening, about 60 positive mutants were produced and a best mutant, Mut H-1287, with improved activity and pH stability was obtained. As compared to wild-type D-pantonohydrolase, Mut H-1287 showed a 10.5-fold higher specific activity; moreover, it could retain 85% of its original activity after incubation under low pH. Gene analysis indicated that the Mut H-1287 had D63H, K118Q, and V241I substitutions. The wild-type and evolved D-pantonohydrolase (Mut H-1287) was purified in three steps. The activities and characteristics of purified wild-type and evolved D-pantonohydrolase were also studied and compared.
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
D-(−)-Pantolactone, 99%