Merck
  • FADS1/2 control lipid metabolism and ferroptosis susceptibility in triple-negative breast cancer.
FADS1/2 control lipid metabolism and ferroptosis susceptibility in triple-negative breast cancer.
EMBO molecular medicine (2024-06-27)
Nicla Lorito, Angela Subbiani, Alfredo Smiriglia, Marina Bacci, Francesca Bonechi, Laura Tronci, Elisabetta Romano, Alessia Corrado, Dario Livio Longo, Marta Iozzo, Luigi Ippolito, Giuseppina Comito, Elisa Giannoni, Icro Meattini, Alexandra Avgustinova, Paola Chiarugi, Angela Bachi, Andrea Morandi
ABSTRACT
Triple-negative breast cancer (TNBC) has limited therapeutic options, is highly metastatic and characterized by early recurrence. Lipid metabolism is generally deregulated in TNBC and might reveal vulnerabilities to be targeted or used as biomarkers with clinical value. Ferroptosis is a type of cell death caused by iron-dependent lipid peroxidation which is facilitated by the presence of polyunsaturated fatty acids (PUFA). Here we identify fatty acid desaturases 1 and 2 (FADS1/2), which are responsible for PUFA biosynthesis, to be highly expressed in a subset of TNBC with a poorer prognosis. Lipidomic analysis, coupled with functional metabolic assays, showed that FADS1/2 high-expressing TNBC are susceptible to ferroptosis-inducing agents and that targeting FADS1/2 by both genetic interference and pharmacological approach renders those tumors ferroptosis-resistant while unbalancing PUFA/MUFA ratio by the supplementation of exogenous PUFA sensitizes resistant tumors to ferroptosis induction. Last, inhibiting lipid droplet (LD) formation and turnover suppresses the buffering capacity of LD and potentiates iron-dependent cell death. These findings have been validated in vitro and in vivo in mouse- and human-derived clinically relevant models and in a retrospective cohort of TNBC patients.
Sigma-Aldrich
ANTI-FADS2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture