Merck
  • Dorsal motor vagal neurons can elicit bradycardia and reduce anxiety-like behavior.
Dorsal motor vagal neurons can elicit bradycardia and reduce anxiety-like behavior.
iScience (2024-02-29)
Misty M Strain, Nicholas J Conley, Lily S Kauffman, Liliana Espinoza, Stephanie Fedorchak, Patricia Castro Martinez, Maisie E Crook, Maira Jalil, Georgia E Hodes, Stephen B G Abbott, Ali D Güler, John N Campbell, Carie R Boychuk
ABSTRACT
Cardiovagal neurons (CVNs) innervate cardiac ganglia through the vagus nerve to control cardiac function. Although the cardioinhibitory role of CVNs in nucleus ambiguus (CVNNA) is well established, the nature and functionality of CVNs in dorsal motor nucleus of the vagus (CVNDMV) is less clear. We therefore aimed to characterize CVNDMV anatomically, physiologically, and functionally. Optogenetically activating cholinergic DMV neurons resulted in robust bradycardia through peripheral muscarinic (parasympathetic) and nicotinic (ganglionic) acetylcholine receptors, but not beta-1-adrenergic (sympathetic) receptors. Retrograde tracing from the cardiac fat pad labeled CVNNA and CVNDMV through the vagus nerve. Using whole-cell patch-clamp, CVNDMV demonstrated greater hyperexcitability and spontaneous action potential firing ex vivo despite similar resting membrane potentials, compared to CVNNA. Chemogenetically activating DMV also caused significant bradycardia with a correlated reduction in anxiety-like behavior. Thus, DMV contains uniquely hyperexcitable CVNs and is capable of cardioinhibition and robust anxiolysis.
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
(−)-Scopolamine methyl bromide, ≥98% (HPLC), powder