Merck
  • BRD4 orchestrates genome folding to promote neural crest differentiation.
BRD4 orchestrates genome folding to promote neural crest differentiation.
Nature genetics (2021-10-07)
Ricardo Linares-Saldana, Wonho Kim, Nikhita A Bolar, Haoyue Zhang, Bailey A Koch-Bojalad, Sora Yoon, Parisha P Shah, Ashley Karnay, Daniel S Park, Jennifer M Luppino, Son C Nguyen, Arun Padmanabhan, Cheryl L Smith, Andrey Poleshko, Qiaohong Wang, Li Li, Deepak Srivastava, Golnaz Vahedi, Gwang Hyeon Eom, Gerd A Blobel, Eric F Joyce, Rajan Jain
ABSTRACT
Higher-order chromatin structure regulates gene expression, and mutations in proteins mediating genome folding underlie developmental disorders known as cohesinopathies. However, the relationship between three-dimensional genome organization and embryonic development remains unclear. Here we define a role for bromodomain-containing protein 4 (BRD4) in genome folding, and leverage it to understand the importance of genome folding in neural crest progenitor differentiation. Brd4 deletion in neural crest results in cohesinopathy-like phenotypes. BRD4 interacts with NIPBL, a cohesin agonist, and BRD4 depletion or loss of the BRD4-NIPBL interaction reduces NIPBL occupancy, suggesting that BRD4 stabilizes NIPBL on chromatin. Chromatin interaction mapping and imaging experiments demonstrate that BRD4 depletion results in compromised genome folding and loop extrusion. Finally, mutation of individual BRD4 amino acids that mediate an interaction with NIPBL impedes neural crest differentiation into smooth muscle. Remarkably, loss of WAPL, a cohesin antagonist, rescues attenuated smooth muscle differentiation resulting from BRD4 loss. Collectively, our data reveal that BRD4 choreographs genome folding and illustrates the relevance of balancing cohesin activity for progenitor differentiation.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
dTAG-13, ≥98% (HPLC)
Sigma-Aldrich
O9-1 Mouse Cranial Neural Crest Cell Line, stably expresses stem cell markers and neural crest markers