Merck
  • High throughput assay for compounds that boost BDNF expression in neurons.
High throughput assay for compounds that boost BDNF expression in neurons.
SLAS discovery : advancing life sciences R & D (2023-02-27)
Guey-Ying Liao, Haifei Xu, Justin Shumate, Louis Scampavia, Timothy Spicer, Baoji Xu
ABSTRACT
Deficiencies in brain-derived neurotrophic factor (BDNF) have been linked to several brain disorders, making compounds that can boost neuronal BDNF synthesis attractive as potential therapeutics. However, a sensitive and quantitative BDNF assay for high-throughput screening (HTS) is still missing. Here we report the generation of a new mouse Bdnf allele, BdnfNLuc, in which the sequence encoding nano luciferase (NLuc) is inserted into the Bdnf locus immediately before the stop codon so that the allele will produce a BDNF-NLuc fusion protein. BDNF-NLuc protein appears to function like BDNF as BdnfNLuc/NLuc homozygous mice grew and behaved almost normally. We were able to establish and optimize cultures of cortical and hippocampal BdnfNLuc/+ neurons isolated from mouse embryos in 384-well plates. We used the cultures as a phenotypic assay to detect the ability of 10 mM KCl to stimulate BDNF synthesis and achieved a reproducible Z' factor > 0.50 for the assay, a measure considered suitable for HTS. We successfully scaled up the assay to screen the 1280-compound LOPAC library (Library of Pharmacologically Active Compounds). The screen identified several BDNF-boosting compounds, one of which is Bay K8644, a L-type voltage-gated calcium channel (L-VGCC) agonist, which was previously shown to stimulate BDNF synthesis. These results indicate that our phenotypic neuronal assay is ready for HTS to identify novel BDNF-boosting compounds.
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Deltamethrin, Synthetic type II pyrethroid. Potent inhibitor of calcineurin. Causes an increase in neurotransmitter release at synapses, and increases intrasynaptosomal Ca2+ levels.
Sigma-Aldrich
LOPAC®1280 (International Version)
Sigma-Aldrich
(±)-Bay K8644, calcium channel agonist
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial