Merck
The use of fetal bovine serum: ethical or scientific problem?
Alternatives to laboratory animals : ATLA (2002-04-25)
Carlo E A Jochems, Jan B F van der Valk, Frans R Stafleu, Vera Baumans
ABSTRACT
Fetal bovine serum (FBS) is a common component of animal cell culture media. It is harvested from bovine fetuses taken from pregnant cows during slaughter. FBS is commonly harvested by means of a cardiac puncture without any form of anaesthesia. Fetuses are probably exposed to pain and/or discomfort, so the current practice of fetal blood harvesting is inhumane. Apart from moral concerns, several scientific and technical problems exist with regard to the use of FBS in cell culture. Efforts should be made to reduce the use of FBS or, preferably, to replace it with synthetic alternatives.
Sigma-Aldrich
FBS Superior, Supplemented FBS, EU approved
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture