Merck
  • Proinflammatory soluble interleukin-15 receptor alpha is increased in rheumatoid arthritis.
Proinflammatory soluble interleukin-15 receptor alpha is increased in rheumatoid arthritis.
Arthritis (2012-08-14)
Ana Cecilia Machado Diaz, Araceli Chico Capote, Celia Aurora Arrieta Aguero, Yunier Rodríguez Alvarez, Diana García Del Barco Herrera, Miguel Estévez Del Toro, Gerardo E Guillen Nieto, Alicia Santos Savio
ABSTRACT
Rheumatoid arthritis (RA) is an autoimmune and inflammatory disease in which many cytokines have been implicated. In particular, IL-15 is a cytokine involved in the inflammatory processes and bone loss. The aim of this study was to investigate the existence in synovial fluid of soluble IL-15Rα, a private receptor subunit for IL-15 which may act as an enhancer of IL-15-induced proinflammatory cytokines. Soluble IL-15Rα was quantified by a newly developed enzyme-linked immunosorbent assay (ELISA) in samples of synovial fluid from patients with RA and osteoarthritis (OA). The levels of IL-15Rα were significantly increased in RA patients compared to OA patients. Also, we studied the presence of membrane-bound IL-15 in cells from synovial fluids, another element necessary to induce pro-inflammatory cytokines through reverse signaling. Interestingly, we found high levels of IL-6 related to high levels of IL-15Rα in RA but not in OA. Thus, our results evidenced presence of IL-15Rα in synovial fluids and suggested that its pro-inflammatory effect could be related to induction of IL-6.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody