Merck
  • Brain-specific suppression of AMPKα2 isoform impairs cognition and hippocampal LTP by PERK-mediated eIF2α phosphorylation.
Brain-specific suppression of AMPKα2 isoform impairs cognition and hippocampal LTP by PERK-mediated eIF2α phosphorylation.
Molecular psychiatry (2020-05-06)
Wenzhong Yang, Xueyan Zhou, Helena R Zimmermann, Tao Ma
ABSTRACT
The AMP-activated protein kinase (AMPK) is a molecular sensor to maintain energy homeostasis. The two isoforms of the AMPK catalytic subunit (AMPKα1 and α2) are both expressed in brains, but their roles in cognition are unknown. We generated conditional knockout mice in which brain AMPKα isoforms are selectively suppressed (AMPKα1/α2 cKO), and determined the isoform-specific effects in mice of either sex on cognition and synaptic plasticity. AMPKα2 cKO but not AMPKα1 cKO displayed impaired cognition and hippocampal late long-term potentiation (L-LTP). Further, AMPKα2 cKO mice exhibited decreased dendritic spine density and abnormal spine morphology in hippocampus. Electron microscope imaging demonstrated reduced postsynaptic density formation and fewer dendritic polyribosomes in hippocampi of AMPKα2 cKO mice. Biochemical studies revealed unexpected findings that repression of AMPKα2 resulted in increased phosphorylation of mRNA translational factor eIF2α and its kinase PERK. Importantly, L-LTP failure and cognitive impairments displayed in AMPKα2 cKO mice were alleviated by suppressing PERK activity pharmacologically or genetically. In summary, we demonstrate here that brain-specific suppression of AMPKα2 isoform impairs cognition and hippocampal LTP by PERK-mediated eIF2α phosphorylation, providing molecular mechanisms linking metabolism, protein synthesis, and cognition.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
PERK Inhibitor I, GSK2606414, GSK2606414 is a cell-permeable, highly potent inhibitor of EIF2AK3/PERK (IC₅₀ = 0.4 nM; [ATP] = 5 µM). Targets PERK in its inactive DFG conformation at the ATP-binding region.
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Anti-Puromycin Sensitive Aminopeptidase Antibody, Chemicon®, from goat