Merck
  • The HIF-1α/p53/miRNA-34a/Klotho axis in retinal pigment epithelial cells promotes subretinal fibrosis and exacerbates choroidal neovascularization.
The HIF-1α/p53/miRNA-34a/Klotho axis in retinal pigment epithelial cells promotes subretinal fibrosis and exacerbates choroidal neovascularization.
Journal of cellular and molecular medicine (2021-01-14)
Laiqing Xie, Ying Wang, Quan Li, Xiaoyan Ji, Yuanyuan Tu, Shu Du, Hui Lou, Xinwei Zeng, Linling Zhu, Ji Zhang, Manhui Zhu
ABSTRACT
Wet age-related macular degeneration (wAMD), characterized by choroidal neovascularization (CNV), is a leading cause of irreversible vision loss among elderly people in developed nations. Subretinal fibrosis, mediated by epithelial-mesenchymal transition (EMT) of retinal pigment epithelium (RPE) cells, leads to unsuccessful anti-vascular endothelial growth factor (VEGF) agent treatments in CNV patients. Under hypoxic conditions, hypoxia-inducible factor-1α (HIF-1α) increases the stability and activation of p53, which activates microRNA-34a (miRNA-34a) transcription to promote fibrosis. Additionally, Klotho is a target gene of miRNA-34a that inhibits fibrosis. This study aimed to explore the role of the HIF-1α/p53/miRNA-34a/Klotho axis in subretinal fibrosis and CNV. Hypoxia-induced HIF-1α promoted p53 stability, phosphorylation and nuclear translocation in ARPE-19 cells (a human RPE cell line). HIF-1α-dependent p53 activation up-regulated miRNA-34a expression in ARPE-19 cells following hypoxia. Moreover, hypoxia-induced p53-dependent miRNA-34a inhibited the expression of Klotho in ARPE-19 cells. Additionally, the HIF-1α/p53/miRNA-34a/Klotho axis facilitated hypoxia-induced EMT in ARPE-19 cells. In vivo, blockade of the HIF-1α/p53/miRNA-34a/Klotho axis alleviated the formation of mouse laser-induced CNV and subretinal fibrosis. In short, the HIF-1α/p53/miRNA-34a/Klotho axis in RPE cells promoted subretinal fibrosis, thus aggravating the formation of CNV.
Sigma-Aldrich
Anti-phospho-p53 (pSer46) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution