Merck
  • In vitro activity of the novel antibacterial agent ibezapolstat (ACX-362E) against Clostridioides difficile.
In vitro activity of the novel antibacterial agent ibezapolstat (ACX-362E) against Clostridioides difficile.
The Journal of antimicrobial chemotherapy (2020-04-15)
Beverly Murray, Cindy Wolfe, Andrea Marra, Chris Pillar, Dean Shinabarger
ABSTRACT
Ibezapolstat (ACX-362E) is the first DNA polymerase IIIC inhibitor undergoing clinical development for the oral treatment of Clostridioides difficile infection (CDI). In this study, the in vitro activity of ibezapolstat was evaluated against a panel of 104 isolates of C. difficile, including those with characterized ribotypes (e.g. 027 and 078) and those producing toxin A or B and was shown to have similar activity to those of comparators against these strains. The overall MIC50/90 (mg/L) for ibezapolstat against evaluated C. difficile was 2/4, compared with 0.5/4 for metronidazole, 1/4 for vancomycin and 0.5/2 for fidaxomicin. In addition, the bactericidal activity of ibezapolstat was evaluated against actively growing C. difficile by determining the MBC against three C. difficile isolates. Time-kill kinetic assays were additionally performed against the three C. difficile isolates, with metronidazole and vancomycin as comparators. The killing of C. difficile by ibezapolstat was observed to occur at concentrations similar to its MIC, as demonstrated by MBC:MIC ratios and reflected in time-kill kinetic assays. This activity highlights the therapeutic potential of ibezapolstat for the treatment of CDI.
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Hemin, from bovine, ≥90%