Merck
  • Immunohistochemical localization of galectin-3 in the granulomatous lesions of paratuberculosis-infected bovine intestine.
Immunohistochemical localization of galectin-3 in the granulomatous lesions of paratuberculosis-infected bovine intestine.
Journal of veterinary science (2009-08-19)
Juyeon Lee, Changjong Moon, Jihoon Kim, Chanwoo Jung, Keun-Hwa Lee, Hong-Gu Joo, Meejung Ahn, Taekyun Shin
ABSTRACT
The presence of galectin-3 was immunohistochemically quantified in bovine intestines infected with paratuberculosis (Johne's disease) to determine whether galectin-3 was involved in the formation of granulation tissue associated with the disease. Mycobacterium avium subsp. paratuberculosis infection was histochemically confirmed using Ziehl-Neelsen staining and molecularly diagnosed through rpoB DNA sequencing. Galectin-3 was detected in the majority of inflammatory cells, possibly macrophages, in the granulomatous lesions within affected tissues, including the ileum. These findings suggest that galectin-3 is associated with the formation of chronic granulation tissues in bovine paratuberculosis, probably through cell adhesion and anti-apoptosis mechanisms.
Sigma-Aldrich
Canada balsam, for microscopy
Sigma-Aldrich
Canada balsam, Mounting medium for microscopy
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission