Merck
  • Analysis of UV ink photoinitiators in packaged food by fast liquid chromatography at sub-ambient temperature coupled to tandem mass spectrometry.
Analysis of UV ink photoinitiators in packaged food by fast liquid chromatography at sub-ambient temperature coupled to tandem mass spectrometry.
Journal of chromatography. A (2010-12-25)
Héctor Gallart-Ayala, Oscar Núñez, Encarnación Moyano, Maria Teresa Galceran
ABSTRACT
A fast method of liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) was developed for the analysis of eleven UV ink photoinitiators in packaged food. Chromatographic separation was achieved in a pentafluorophenylpropyl (PFPP) column at 5°C and acetonitrile:25 mM formic acid-ammonium formate (pH 2.7) in gradient elution. To reduce sample treatment, a QuEChERS (quick, easy, cheap, effective, rugged and safe) method for the extraction and clean-up of UV photoinitiators in packaged foods was evaluated. Triple quadrupole working in H-SRM on Q1 mode was used for both quantitation and confirmation purposes and the most intense and selective transitions were chosen. Quality parameters of the developed QuEChERS LC-MS/MS method were established and applied for the analysis of photoinitiators in food packaged at ng kg(-1) levels.
Product Description
Supelco
2-Isopropylthioxanthone, PESTANAL®, analytical standard
Supelco
4-Isopropylthioxanthone, PESTANAL®, analytical standard