Merck
  • Use of copper and insulin-resistance to accelerate cognitive deficits and synaptic protein loss in a rat Abeta-infusion Alzheimer's disease model.
Use of copper and insulin-resistance to accelerate cognitive deficits and synaptic protein loss in a rat Abeta-infusion Alzheimer's disease model.
Journal of Alzheimer's disease : JAD (2008-12-20)
Aynun N Begum, Fusheng Yang, Edmond Teng, Shuxin Hu, Mychica R Jones, Emily R Rosario, Walter Beech, Beverly Hudspeth, Oliver J Ubeda, Greg M Cole, Sally A Frautschy
ABSTRACT
The rat amyloid-beta (Abeta) intracerebroventricular infusion can model aspects of Alzheimer's disease (AD) and has predicted efficacy of therapies such as ibuprofen and curcumin in transgenic mouse models. High density lipoprotein (HDL), a normal plasma carrier of Abeta, is used to attenuate Abeta aggregation within the pump, causing Abeta-dependent toxicity and cognitive deficits within 3 months. Our goal was to identify factors that might accelerate onset of Abeta-dependent deficits to improve efficiency and cost-effectiveness of model. We focused on: 1) optimizing HDL-Abeta preparation for maximal toxicity; 2) evaluating the role of copper, a factor typically in water that can impact oligomer stability; and 3) determining impact of insulin resistance (type II diabetes), a risk factor for AD. In vitro studies were performed to determine doses of copper and methods of Abeta-HDL preparation that maximized toxicity. These preparations when infused resulted in earlier onset of cognitive deficits within 6 weeks post-infusion. Induction of insulin resistance did not exacerbate Abeta-dependent cognitive deficits, but did exacerbate synaptic protein loss. In summary, the newly described in vivo infusion model may be useful cost-effective method for screening for new therapeutic drugs for AD.
Sigma-Aldrich
Amyloid β 1-42 rat, ≥90% (HPLC)
Sigma-Aldrich
Amyloid β 1-40 rat, ≥95% (HPLC)
Sigma-Aldrich
Lipoproteins, High Density, Human Plasma
Sigma-Aldrich
Amyloid β 1-16 rat