Merck
  • Gal-3BP Negatively Regulates NF-κB Signaling by Inhibiting the Activation of TAK1.
Gal-3BP Negatively Regulates NF-κB Signaling by Inhibiting the Activation of TAK1.
Frontiers in immunology (2019-08-14)
Chang-Soo Hong, Mi-Ra Park, Eun-Gene Sun, Wonyoung Choi, Jun-Eul Hwang, Woo-Kyun Bae, Joon Haeng Rhee, Sang-Hee Cho, Ik-Joo Chung
ABSTRACT
Galectin-3-binding protein (Gal-3BP) is a member of the family of scavenger receptor cysteine-rich (SRCR) domain-containing proteins, which are associated with the immune system. However, the functional roles and signaling mechanisms of Gal-3BP in host defense and the immune response remain largely unknown. Here, we identified cellular Gal-3BP as a negative regulator of NF-κB activation and proinflammatory cytokine production in lipopolysaccharide (LPS)-stimulated murine embryonic fibroblasts (MEFs). Furthermore, cellular Gal-3BP interacted with transforming growth factor β-activated kinase 1 (TAK1), a crucial mediator of NF-κB activation in response to cellular stress. Gal-3BP inhibited the phosphorylation of TAK1, leading to suppression of its kinase activity and reduced protein stability. In vivo we found that Lgals3BP deficiency in mice enhanced LPS-induced proinflammatory cytokine release and rendered mice more sensitive to LPS-induced endotoxin shock. Overall, these results suggest that Gal-3BP is a novel suppressor of TAK1-dependent NF-κB activation that may have potential in the prevention and treatment of inflammatory diseases.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-TAK1 (Ser412) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)