Merck
  • Immature Low-Density Neutrophils Exhibit Metabolic Flexibility that Facilitates Breast Cancer Liver Metastasis.
Immature Low-Density Neutrophils Exhibit Metabolic Flexibility that Facilitates Breast Cancer Liver Metastasis.
Cell reports (2019-06-27)
Brian E Hsu, Sébastien Tabariès, Radia M Johnson, Sylvia Andrzejewski, Julien Senecal, Camille Lehuédé, Matthew G Annis, Eric H Ma, Sandra Völs, LeeAnn Ramsay, Remi Froment, Anie Monast, Ian R Watson, Zvi Granot, Russell G Jones, Julie St-Pierre, Peter M Siegel
ABSTRACT
Neutrophils are phenotypically heterogeneous and exert either anti- or pro-metastatic functions. We show that cancer-cell-derived G-CSF is necessary, but not sufficient, to mobilize immature low-density neutrophils (iLDNs) that promote liver metastasis. In contrast, mature high-density neutrophils inhibit the formation of liver metastases. Transcriptomic and metabolomic analyses of high- and low-density neutrophils reveal engagement of numerous metabolic pathways specifically in low-density neutrophils. iLDNs exhibit enhanced global bioenergetic capacity, through their ability to engage mitochondrial-dependent ATP production, and remain capable of executing pro-metastatic neutrophil functions, including NETosis, under nutrient-deprived conditions. We demonstrate that NETosis is an important neutrophil function that promotes breast cancer liver metastasis. iLDNs rely on the catabolism of glutamate and proline to support mitochondrial-dependent metabolism in the absence of glucose, which enables sustained NETosis. These data reveal that distinct pro-metastatic neutrophil populations exhibit a high degree of metabolic flexibility, which facilitates the formation of liver metastases.
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
BPTES, ≥95% (HPLC)
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Roche
DNase I recombinant, grade I, from bovine pancreas, expressed in Pichia pastoris