
Valnemulin hydrochloride

Empirical Formula (Hill Notation):
C31H52N2O5S · HCl
CAS Number:
Molecular Weight:
601.28
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3