Skip to Content
Merck
Trimethyl(trifluoromethyl)silane 99%

Trimethyl(trifluoromethyl)silane

Trimethyl(trifluoromethyl)silane 99%
Synonym(s):
(Trifluoromethyl)trimethylsilane, Ruppert’s Reagent, TFMTMS
Linear Formula:
(CH3)3SiCF3
CAS Number:
Molecular Weight:
142.19

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing