
Thrombopoietin human

Synonym(s):
MGDF, Megakaryocyte colony-stimulating factor, TPO, c-MPL ligand
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing