Skip to Content
EE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
tert
-Butylamine
Synonym(s):
2-Amino-2-methylpropane, tert-Butylamine
Linear Formula:
(CH
3
)
3
CNH
2
CAS Number:
75-64-9
Molecular Weight:
73.14
Products
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
B89205
tert
-Butylamine,
98%
Expand
View Pricing
391433
tert
-Butylamine,
≥99.5%
Expand
View Pricing
8.01546
tert
-Butylamine,
for synthesis
Expand
View Pricing