
Rosuvastatin impurity G

Synonym(s):
(3S,5R,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid
Empirical Formula (Hill Notation):
C22H28FN3O6S
CAS Number:
Molecular Weight:
481.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing