
Riboflavin sodium phosphate for peak identification

Synonym(s):
Riboflavin 5′-monophosphate sodium salt, Riboflavin Sodium phosphate
Empirical Formula (Hill Notation):
C17H20N4NaO9P
CAS Number:
Molecular Weight:
478.33
Products
Brand
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing