
Rabeprazole Related Compound C

Synonym(s):
2-Mercaptobenzimidazole, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol
Empirical Formula (Hill Notation):
C7H6N2S
CAS Number:
Molecular Weight:
150.20
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2