
Rabeprazole Related Compound B

Synonym(s):
Rabeprazole N-oxide, 2-[[4-(3-Methoxypropoxy)-3-methyl-1-oxypyridin-2-yl]methylsulfinyl]-1H-benzimidazole
Empirical Formula (Hill Notation):
C18H21N3O4S
CAS Number:
Molecular Weight:
375.44
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2