
Pyrithiobac

Synonym(s):
2-Chloro-6-[(4,6-dimethoxy-2-pyrimidinyl)thio]benzoic acid
Empirical Formula (Hill Notation):
C13H11ClN2O4S
CAS Number:
Molecular Weight:
326.76
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing