
Pyraflufen-ethyl

Synonym(s):
Ethyl [2-chloro-5-(4-chloro-5-difluoromethoxy-1-methylpyrazol-3-yl)-4-fluorophenoxy]acetate
Empirical Formula (Hill Notation):
C15H13Cl2F3N2O4
CAS Number:
Molecular Weight:
413.18
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2