
Poly(ethylene glycol) methyl ether thiol

Synonym(s):
Methoxy PEG thiol, Methoxypolyethylene glycol thiol, PEG thiol, mPEG thiol
Linear Formula:
CH3O(CH2CH2O)nCH2CH2SH
Products
Brand
Melting Point (°C)
Physical Form
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing