
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate

Linear Formula:
C3H3O[C2H4O]x[C3H6O]y[C2H4O]zC3H3O2
Products
Brand
Color
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing