
Pidolic acid

Synonym(s):
L-Pyroglutamic acid, (S)-(−)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid
Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing