
Parathion

Synonym(s):
O,O-Diethyl O-(4-nitrophenyl) phosphorothioate, Ethylparathione, Parathion-ethyl
Empirical Formula (Hill Notation):
C10H14NO5PS
CAS Number:
Molecular Weight:
291.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2