
Mollugin

Synonym(s):
6-Hydroxy-2,2-dimethyl-2H-naphtho(1,2-b)pyran-5-carboxylic acid methyl ester, Rubimaillin
Empirical Formula (Hill Notation):
C17H16O4
CAS Number:
Molecular Weight:
284.31
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing