
Methylnaltrexone bromide

Synonym(s):
17-(Cyclopropylmethyl)-4,5α-epoxy-3,14-dihydroxy-17-methyl-6-oxomorphinanium bromide, MNTX, Methylnaltrexonium, Mrz-2663, N-Methylnaltrexone, Naltrexone MB, Quaternary ammonium naltrexone
Empirical Formula (Hill Notation):
C21H26NO4 · Br
CAS Number:
Molecular Weight:
436.34
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade