
Magnesium sulfate

Synonym(s):
Magnesium sulfate heptahydrate, Epsom salts
Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2