
Levalbuterol Related Compound E

Synonym(s):
4-[2-(tert-Butylamino)-1-hydroxyethyl]-2-(ethoxymethyl)phenol, α-{[(1,1-Dimethylethyl)amino]methyl}-3-(ethoxymethyl)-4-hydroxybenzenemethanol, Albuterol monoethyl ether
Empirical Formula (Hill Notation):
C15H25NO3
CAS Number:
Molecular Weight:
267.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2