
Lectin from Phaseolus vulgaris (red kidney bean)

Synonym(s):
Phaseolus vulgaris agglutinin, PHA
Products
Brand
Biological Source
Physical Form
Product Line
Chemical Composition
Quality Segment
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing