Skip to Content
Merck
L-Cysteine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Cysteine

L-Cysteine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(R)-2-Amino-3-mercaptopropionic acid, L-Cysteine
Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Technique